N,N-Dimethylacetamide, 99+%, for spectroscopy
N,N-Dimethylacetamide, 99+%, for spectroscopy
N,N-Dimethylacetamide, 99+%, for spectroscopy
N,N-Dimethylacetamide, 99+%, for spectroscopy
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, for spectroscopy
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass Bottle
Catalog number FSA167782500
Price (MYR)
343.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
343.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
Melting Point-20.0°C
Absorbance0.01 max. at 360nm, 0.03 max. at 320nm, 0.05 max. at 310nm, 0.15 max. at 290nm, 0.3 max. at 280nm, 1 max. at 270nm
CAS127-19-5
CAS Min %99.0
View more
For UV/VIS spectroscopy
RUO – Research Use Only