1-Butanol, 99+%, for spectroscopy
1-Butanol, 99+%, for spectroscopy
1-Butanol, 99+%, for spectroscopy
1-Butanol, 99+%, for spectroscopy
Thermo Scientific Chemicals
1-Butanol, 99+%, for spectroscopy
CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass Bottle
Catalog number FSA167695000
Price (MYR)
331.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
331.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Butanol
Melting Point-89.0°C
Absorbance0.097 max. at 238nm, 1 max. at 205nm, 0.4 max. at 220nm, 0.1 max. at 240nm, 0.04 max. at 260nm, 0.02 max. at 280nm, 0.01 max. 330nm, 0.602 max. at 210nm, 0.046 max. at 245nm, 0.009 max. at 270nm, (1 cm cell vs HPLC-grade water), (1 cm cell vs HPLC-grade water) at 0nm
CAS71-36-3
CAS Min %99.0
View more
For UV/VIS spectroscopy
RUO – Research Use Only