4-Methylcyclohexanol, 99%, mixture of cis and trans
4-Methylcyclohexanol, 99%, mixture of cis and trans
4-Methylcyclohexanol, 99%, mixture of cis and trans
4-Methylcyclohexanol, 99%, mixture of cis and trans
Thermo Scientific Chemicals
4-Methylcyclohexanol, 99%, mixture of cis and trans
CAS: 589-91-3 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA166902500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Methylcyclohexanol
Name Note99%
CAS589-91-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning