1-Bromooctane, 99%
1-Bromooctane, 99%
1-Bromooctane, 99%
Thermo Scientific Chemicals
1-Bromooctane, 99%
CAS: 111-83-1 | C8H17Br | 193.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA166815000
Price (MYR)
505.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
505.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromooctane
CAS111-83-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.