2-Methyl-2-butanol, 99%, pure
2-Methyl-2-butanol, 99%, pure
2-Methyl-2-butanol, 99%, pure
2-Methyl-2-butanol, 99%, pure
Thermo Scientific Chemicals
2-Methyl-2-butanol, 99%, pure
CAS: 75-85-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA166620025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-2-butanol
Name Note99%
CAS75-85-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful in contact with skin.
May cause drowsiness or d