4-Chlorophenyl isocyanate, 98%
4-Chlorophenyl isocyanate, 98%
4-Chlorophenyl isocyanate, 98%
4-Chlorophenyl isocyanate, 98%
Thermo Scientific Chemicals
4-Chlorophenyl isocyanate, 98%
CAS: 104-12-1 | C7H4ClNO | 153.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA166565000
Price (MYR)
1,102.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,102.00
EA
Add to cart
Specifications
Chemical Name or Material4-Chlorophenyl isocyanate
Melting Point29.0°C to 31.0°C
CAS104-12-1
CAS Min %97.5
ColorWhite to Yellow