Flufenamic acid, 97%
Flufenamic acid, 97%
Flufenamic acid, 97%
Flufenamic acid, 97%
Thermo Scientific Chemicals
Flufenamic acid, 97%
CAS: 530-78-9 | C14H10F3NO2 | 281.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA165920500
Price (MYR)
1,097.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,097.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFlufenamic acid
Name Note97%
CAS530-78-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement