Ninhydrin, spectrophotometric grade
Ninhydrin, spectrophotometric grade
Ninhydrin, spectrophotometric grade
Ninhydrin, spectrophotometric grade
Thermo Scientific Chemicals
Ninhydrin, spectrophotometric grade
CAS: 485-47-2 | C9H6O4 | 178.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA165875000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNinhydrin
Melting Point250.0°C
CAS485-47-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement: