DL-1,4-Dithiothreitol, 99%, for biochemistry
DL-1,4-Dithiothreitol, 99%, for biochemistry
DL-1,4-Dithiothreitol, 99%, for biochemistry
DL-1,4-Dithiothreitol, 99%, for biochemistry
Thermo Scientific Chemicals
DL-1,4-Dithiothreitol, 99%, for biochemistry
1,4-Dithiothreitol, CAS # 3483-12-3, is a reducing agent commonly used to reduce disulphide bonds in protein studies, such as western blot SDS-page electrophoresis. | CAS: 3483-12-03 | C4H10O2S2 | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 g
5 g
25 g
Catalog number FSA165685000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-1, 4-Dithiothreitol
CAS3483-12-03
Absorbance(1M soln. in water), (1 M soln. in water) at 0nm, 0.035 max. at 500nm
Health Hazard 1Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Health Hazard 2Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, May form combustible dust concentrations in air, Causes skin irritation
View more
General description
  • 1,4-Dithiothreitol (DTT or Cleland's reagent) reduces disulfide bonds in proteins, leading to loss of native conformation
  • DTT is not stable in solution and should be prepared in small aliquots prior to use; the use of EDTA in the solution can increase the half-life of DTT
  • Typically used at a final concentration of 0.1M for protein stabilization; recommended to create a 1M stock
Applications
  • Protects thiol groups from oxidation and acts as a chemical chelator in solution; maintains proteins in the denatured state
  • Common applications include SDS-page electrophoresis and protein isolation
RUO – Research Use Only