5alpha-Cholestane, 98+%
5alpha-Cholestane, 98+%
5alpha-Cholestane, 98+%
5alpha-Cholestane, 98+%
Thermo Scientific Chemicals
5alpha-Cholestane, 98+%
CAS: 481-21-0 | C27H48 | 372.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA165602500
Price (MYR)
597.00
BOT
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
597.00
BOT
Add to cart
Specifications
Chemical Name or Material5α-Cholestane
CAS481-21-0
CAS Min %98.0
CAS Max %100.0
Melting Point78°C to 81°C