Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans
Thermo Scientific Chemicals
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans
CAS: 19438-60-9 | C9H12O3 | 168.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA165395000
Price (MYR)
806.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
806.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexahydro-4-methylphthalic anhydride
Name Note98%
CAS19438-60-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.