2,5-Dihydroxybenzoic acid, 99%
2,5-Dihydroxybenzoic acid, 99%
2,5-Dihydroxybenzoic acid, 99%
2,5-Dihydroxybenzoic acid, 99%
Thermo Scientific Chemicals
2,5-Dihydroxybenzoic acid, 99%
CAS: 490-79-9 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA165200500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 5-Dihydroxybenzoic acid
CAS490-79-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0