Acrylamide, 99+%, for biochemistry, electrophoresis grade
Acrylamide, 99+%, for biochemistry, electrophoresis grade
Acrylamide, 99+%, for biochemistry, electrophoresis grade
Acrylamide, 99+%, for biochemistry, electrophoresis grade
Thermo Scientific Chemicals
Acrylamide, 99+%, for biochemistry, electrophoresis grade
Acrylamide, CAS # 79-06-1, is an organic amide compound used as a monomer in the synthesis of polyacrylamides. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic Bottle
Catalog number FSA164855000
Price (MYR)
656.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
656.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcrylamide
Name Note99+%
Physical FormCrystals or Crystalline Powder
Acrylic Acid0.002% max.
CAS79-06-1
View more
General Description
• Acrylamide is an organic amide compound mainly used to as a monomer in the synthesis of polyacrylamide
• Acrylamide is formed in foods containing carbohydrates in its composition, mainly when these foods are fried, grilled or baked
Applications
• In laboratory experiments acrylamide participates in chemical reactions as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide compounds
• For molecular usage, this compound can be used as a carrier and is involved in the precipitation of small amounts of DNA
• In molecular experiments it is used in gel electrophoresis, including western blot, and SDS-PAGE, to separate proteins and macromolecules based on their size and charge
RUO – Research Use Only
General References:
  1. Pelucchi, C.; Bosetti, C.; Galeone, C.; La Vecchia, C. Dietary acrylamide and cancer risk: An updated meta-analysis. Int. J. Cancer 2015, 136 (12), 2912-2922.
  2. Vaclavik, L.; Capuano, E.; Gökmen, V.; Hajslova, J. Prediction of acrylamide formation in biscuits based on fingerprint data generated by ambient ionization mass spectrometry employing direct analysis in real time (DART) ion source. Food Chem. 2015, 173, 290-297.