Acrylamide, 98.5%, extra pure
Acrylamide, 98.5%, extra pure
Acrylamide, 98.5%, extra pure
Acrylamide, 98.5%, extra pure
Thermo Scientific Chemicals
Acrylamide, 98.5%, extra pure
Acrylamide, CAS # 79-06-1, is an organic amide compound used as a monomer in the synthesis of polyacrylamides. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA164835000
Price (MYR)
651.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
651.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcrylamide
Physical FormCrystals or Crystalline Powder
CAS79-06-1
Boiling Point125.0°C (25.0 mmHg)
Health Hazard 1Danger
View more
General Description
  • Acrylamide is an organic amide compound mainly used to as a monomer in the synthesis of polyacrylamide
  • Acrylamide is formed in foods containing carbohydrates in its composition, mainly when these foods are fried, grilled or baked
Applications
  • In laboratory experiments acrylamide participates in chemical reactions as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide compounds
  • For molecular usage, this compound can be used as a carrier and is involved in the precipitation of small amounts of DNA
  • In molecular experiments it is used in gel electrophoresis, including western blot, and SDS-PAGE, to separate proteins and macromolecules based on their size and charge
RUO – Research Use Only