Thiobenzamide, 95%
Thiobenzamide, 95%
Thiobenzamide, 95%
Thermo Scientific Chemicals
Thiobenzamide, 95%
CAS: 2227-79-4 | C7H7NS | 137.20 g/mol
Have Questions?
Catalog NumberQuantity
FSA16473010010 g
Catalog number FSA164730100
Price (MYR)
300.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
300.00
EA
Add to cart
Specifications
Chemical Name or MaterialThiobenzamide
Name Note95%
CAS2227-79-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: