2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
Thermo Scientific Chemicals
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
CAS: 77-99-6 | C6H14O3 | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA164655000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Ethyl-2-(hydroxymethyl)-1, 3-propanediol
Acidity0.002% max. (as formic acid)
CAS77-99-6
CAS Min %97.5
CAS Max %100.0