1,3,5-Benzenetricarboxylic acid chloride, 98%
1,3,5-Benzenetricarboxylic acid chloride, 98%
1,3,5-Benzenetricarboxylic acid chloride, 98%
1,3,5-Benzenetricarboxylic acid chloride, 98%
Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid chloride, 98%
CAS: 4422-95-1 | C9H3Cl3O3 | 265.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA164375000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 3, 5-Benzenetricarboxylic acid chloride
CAS4422-95-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0