alpha-Pinene, 97%, stabilized with alpha-Tocopherol
alpha-Pinene, 97%, stabilized with alpha-Tocopherol
alpha-Pinene, 97%, stabilized with alpha-Tocopherol
alpha-Pinene, 97%, stabilized with alpha-Tocopherol
Thermo Scientific Chemicals
alpha-Pinene, 97%, stabilized with alpha-Tocopherol
CAS: 80-56-8 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number FSA164362500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialα-Pinene
Name Note97%
CAS80-56-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger