Heptanoic acid, 98%
Heptanoic acid, 98%
Heptanoic acid, 98%
Thermo Scientific Chemicals
Heptanoic acid, 98%
CAS: 111-14-8 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA164172500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHeptanoic acid
Name Note98%
CAS111-14-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement