2-Hydroxy-5-methoxybenzaldehyde, 98%
2-Hydroxy-5-methoxybenzaldehyde, 98%
2-Hydroxy-5-methoxybenzaldehyde, 98%
2-Hydroxy-5-methoxybenzaldehyde, 98%
Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%
CAS: 672-13-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Packaging:
Glass bottle
Catalog number FSA164160100
Price (MYR)
724.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
724.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Hydroxy-5-methoxybenzaldehyde
CAS672-13-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0