Bathocuproinedisulfonic acid, disodium salt hydrate, 97%
Bathocuproinedisulfonic acid, disodium salt hydrate, 97%
Thermo Scientific Chemicals
Bathocuproinedisulfonic acid, disodium salt hydrate, 97%
Bathocuproinedisulfonic acid, disodium salt hydrate, 97%, CAS Number-1257642-74-2| C26H18N2Na2O6S2 xH2O | 564.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA164060010
Price (MYR)
844.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
844.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBathocuproinedisulfonic acid, disodium salt hydrate
CAS1257642-74-2
Melting Point300.0°C
EINECS Number258-111-7
Assay Percent Rangedisulfonate) (HPLC)