Benzyltriethylammonium chloride, 98%
Benzyltriethylammonium chloride, 98%
Benzyltriethylammonium chloride, 98%
Benzyltriethylammonium chloride, 98%
Thermo Scientific Chemicals
Benzyltriethylammonium chloride, 98%
CAS: 56-37-1 | C13H22ClN | 227.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA164021000
Price (MYR)
250.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
250.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyltriethylammonium chloride
Name Note98%
CAS56-37-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
View more
Phase transfer catalyst
RUO – Research Use Only