7-Amino-1,3-naphthalenedisulfonic acid, Tech.
7-Amino-1,3-naphthalenedisulfonic acid, Tech.
7-Amino-1,3-naphthalenedisulfonic acid, Tech.
7-Amino-1,3-naphthalenedisulfonic acid, Tech.
Thermo Scientific Chemicals
7-Amino-1,3-naphthalenedisulfonic acid, Tech.
CAS: 86-65-7 | C10H9NO6S2 | 303.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA163961000
Price (MYR)
338.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
338.00
BOT
Add to cart
Specifications
Chemical Name or Material7-Amino-1, 3-naphthalenedisulfonic acid
Name Notetech.
CAS86-65-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: