2-Methyl-4-nitroaniline, 99%
2-Methyl-4-nitroaniline, 99%
2-Methyl-4-nitroaniline, 99%
2-Methyl-4-nitroaniline, 99%
Thermo Scientific Chemicals
2-Methyl-4-nitroaniline, 99%
CAS: 99-52-5 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA163950050
Price (MYR)
225.00
BOT
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
225.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Methyl-4-nitroaniline
Name Note99%
CAS99-52-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger