1-Ethyl-2-pyrrolidinone, 98%
1-Ethyl-2-pyrrolidinone, 98%
1-Ethyl-2-pyrrolidinone, 98%
1-Ethyl-2-pyrrolidinone, 98%
Thermo Scientific Chemicals
1-Ethyl-2-pyrrolidinone, 98%
CAS: 2687-91-4 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA163931000
Price (MYR)
762.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
762.00
EA
Add to cart
Specifications
Chemical Name or Material1-Ethyl-2-pyrrolidinone
CAS2687-91-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0