3-Cyanobenzaldehyde, 99%
3-Cyanobenzaldehyde, 99%
3-Cyanobenzaldehyde, 99%
3-Cyanobenzaldehyde, 99%
Thermo Scientific Chemicals
3-Cyanobenzaldehyde, 99%
CAS: 24964-64-5 | C8H5NO | 131.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA163900050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Cyanobenzaldehyde
Name Note99%
CAS24964-64-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning