Silicone oil, for melting point and boiling point apparatuses
Silicone oil, for melting point and boiling point apparatuses
Silicone oil, for melting point and boiling point apparatuses
Silicone oil, for melting point and boiling point apparatuses
Thermo Scientific Chemicals
Silicone oil, for melting point and boiling point apparatuses
CAS: 63148-62-9 | (C2H6OSi)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number FSA163855000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSilicone oil
Melting Point-50.0°C
Name NoteFor melting point and boiling point apparatuses
CAS63148-62-9