2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
CAS: 6976-37-0 | C8H19NO5 | 209.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA163825000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Name Note98%
CAS6976-37-0
Absorbance(0.1M in H2O)
CAS Min %97.5