Quinine sulfate dihydrate, 99+%
Quinine sulfate dihydrate, 99+%
Quinine sulfate dihydrate, 99+%
Quinine sulfate dihydrate, 99+%
Thermo Scientific Chemicals
Quinine sulfate dihydrate, 99+%
CAS: 6119-70-6 | C40H54N4O10S | 782.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA163710500
Price (MYR)
1,190.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,190.00
BOT
Add to cart
Specifications
Chemical Name or MaterialQuinine sulfate dihydrate
Name Note99+%
CAS6119-70-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement