Quinine, 98+%, anhydrous
Quinine, 98+%, anhydrous
Quinine, 98+%, anhydrous
Quinine, 98+%, anhydrous
Thermo Scientific Chemicals
Quinine, 98+%, anhydrous
CAS: 130-95-0 | C20H24N2O2 | 324.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA163702500
Price (MYR)
5,479.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,479.00
EA
Add to cart
Specifications
Chemical Name or MaterialQuinine
CAS130-95-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If s