Adenine sulfate, 98%, synthetic
Adenine sulfate, 98%, synthetic
Adenine sulfate, 98%, synthetic
Adenine sulfate, 98%, synthetic
Thermo Scientific Chemicals
Adenine sulfate, 98%, synthetic
CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA163630250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAdenine sulfate
Name Note98%
CAS321-30-2
Absorbance0.74 to 0.78 (A250/A260), 0.34 to 0.38 (A280/A260)
CAS Min %97.5