1-Bromodecane, 98%
1-Bromodecane, 98%
1-Bromodecane, 98%
Thermo Scientific Chemicals
1-Bromodecane, 98%
CAS: 112-29-8 | C10H21Br | 221.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA163605000
Price (MYR)
603.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
603.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromodecane
Name Note98%
CAS112-29-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning