1,3-Dihydroxynaphthalene, 99+%
1,3-Dihydroxynaphthalene, 99+%
1,3-Dihydroxynaphthalene, 99+%
1,3-Dihydroxynaphthalene, 99+%
Thermo Scientific Chemicals
1,3-Dihydroxynaphthalene, 99+%
CAS: 132-86-5 | C10H8O2 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA163340250
Price (MYR)
2,656.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,656.00
EA
Add to cart
Specifications
Chemical Name or Material1, 3-Dihydroxynaphthalene
CAS132-86-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for