alpha-Bromo-o-tolunitrile, 98%
alpha-Bromo-o-tolunitrile, 98%
alpha-Bromo-o-tolunitrile, 98%
alpha-Bromo-o-tolunitrile, 98%
Thermo Scientific Chemicals
alpha-Bromo-o-tolunitrile, 98%
CAS: 22115-41-9 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA163130250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialα-Bromo-o-tolunitrile
CAS22115-41-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0