Chlorodimethylsilane, 97%
Chlorodimethylsilane, 97%
Chlorodimethylsilane, 97%
Chlorodimethylsilane, 97%
Thermo Scientific Chemicals
Chlorodimethylsilane, 97%
CAS: 1066-35-9 | C2H6ClSi | 93.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA162842500
Price (MYR)
1,601.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,601.00
EA
Add to cart
Specifications
Chemical Name or Materialchlorodimethylsilane
CAS1066-35-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Extremely flammable liquid and vapor.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
View more
Reduction reagent
RUO – Research Use Only