3,4-Dimethoxybenzaldehyde, 99+%
3,4-Dimethoxybenzaldehyde, 99+%
3,4-Dimethoxybenzaldehyde, 99+%
3,4-Dimethoxybenzaldehyde, 99+%
Thermo Scientific Chemicals
3,4-Dimethoxybenzaldehyde, 99+%
CAS: 120-14-9 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA162615000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 4-Dimethoxybenzaldehyde
Name Note99+%
CAS120-14-9
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning