Diglycolic acid, 98%
Diglycolic acid, 98%
Diglycolic acid, 98%
Diglycolic acid, 98%
Thermo Scientific Chemicals
Diglycolic acid, 98%
CAS: 110-99-6 | C4H6O5 | 134.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA162261000
Price (MYR)
476.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
476.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiglycolic acid
Name Note98%
CAS110-99-6
Melting Point141.0°C to 145.0°C
Health Hazard 1GHS Signal Word: Danger