1-Dimethylamino-2-propyne, 97%
1-Dimethylamino-2-propyne, 97%
1-Dimethylamino-2-propyne, 97%
1-Dimethylamino-2-propyne, 97%
Thermo Scientific Chemicals
1-Dimethylamino-2-propyne, 97%
CAS: 7223-38-3 | C5H9N | 83.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA162250050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Dimethylamino-2-propyne
Name Note97%
CAS7223-38-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger