N,N-Dimethylformamide diethyl acetal, 95%
N,N-Dimethylformamide diethyl acetal, 95%
N,N-Dimethylformamide diethyl acetal, 95%
N,N-Dimethylformamide diethyl acetal, 95%
Thermo Scientific Chemicals
N,N-Dimethylformamide diethyl acetal, 95%
CAS: 1188-33-6 | C7H17NO2 | 147.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number FSA162080500
Price (MYR)
1,327.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,327.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide diethyl acetal
CAS1188-33-6
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Alkylation reagent
RUO – Research Use Only