1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
Thermo Scientific Chemicals
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, C5H12N2S2, CAS Number-5108-96-3 | CAS: 5108-96-3 | C5H12N2S2 | 164.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA162055000
Price (MYR)
4,739.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,739.00
EA
Add to cart
Specifications
Chemical Name or Material1-Pyrrolidinecarbodithioic acid, ammonium salt
CAS5108-96-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0