Chlorosulfonyl isocyanate, 98+%
Chlorosulfonyl isocyanate, 98+%
Chlorosulfonyl isocyanate, 98+%
Chlorosulfonyl isocyanate, 98+%
Thermo Scientific Chemicals
Chlorosulfonyl isocyanate, 98+%
CAS: 1189-71-5 | CClNO3S | 141.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
50 mL
1 L
Packaging:
Glass bottle
Catalog number FSA162042500
Price (MYR)
2,257.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,257.00
BOT
Add to cart
Specifications
Chemical Name or MaterialChlorosulfonyl isocyanate
CAS1189-71-5
Health Hazard 1GHS Signal Word: Danger
Melting Point-44.0°C
ColorWhite
View more
Alkylation reagent, C-X bond formation reagent
RUO – Research Use Only