2,2,2-Trichloroethyl chloroformate, 97%
2,2,2-Trichloroethyl chloroformate, 97%
2,2,2-Trichloroethyl chloroformate, 97%
2,2,2-Trichloroethyl chloroformate, 97%
Thermo Scientific Chemicals
2,2,2-Trichloroethyl chloroformate, 97%
CAS: 17341-93-4 | C3H2Cl4O2 | 211.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA161811000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 2, 2-Trichloroethyl chloroformate
Name Note96%
CAS17341-93-4
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger
View more
Coupling reagent
RUO – Research Use Only