Cytosine, 99+%
Cytosine, 99+%
Cytosine, 99+%
Thermo Scientific Chemicals
Cytosine, 99+%
CAS: 71-30-7 | C4H5N3O | 111.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA161760050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCytosine
CAS71-30-7
Melting Point>300.0°C
ColorWhite to Yellow
EINECS Number200-749-5