4-Nitroimidazole, 98%
4-Nitroimidazole, 98%
4-Nitroimidazole, 98%
4-Nitroimidazole, 98%
Thermo Scientific Chemicals
4-Nitroimidazole, 98%
CAS: 3034-38-6 | C3H3N3O2 | 113.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
View all
Plastic Bottle
Catalog number FSA161605000
Price (MYR)
3,721.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
3,721.00
EA
Add to cart
Specifications
Chemical Name or Material4-Nitroimidazole
CAS3034-38-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0