D(-)-Arabinose, 99+%
D(-)-Arabinose, 99+%
D(-)-Arabinose, 99+%
D(-)-Arabinose, 99+%
Thermo Scientific Chemicals
D(-)-Arabinose, 99+%
CAS: 28697-53-2 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA161450250
Price (MYR)
384.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
384.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD(-)-Arabinose
Name Notep.a.
CAS28697-53-2
Melting Point152.0°C to 160.0°C
Infrared SpectrumAuthentic