4-Methoxystyrene, 96%, stabilized
4-Methoxystyrene, 96%, stabilized
4-Methoxystyrene, 96%, stabilized
4-Methoxystyrene, 96%, stabilized
Thermo Scientific Chemicals
4-Methoxystyrene, 96%, stabilized
CAS: 637-69-4 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA161390250
Price (MYR)
1,080.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,080.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Methoxystyrene
Name Note96%
CAS637-69-4
CAS Min %95.0
CAS Max %100.0