1,4-Dibromobutane, 99%
1,4-Dibromobutane, 99%
1,4-Dibromobutane, 99%
1,4-Dibromobutane, 99%
Thermo Scientific Chemicals
1,4-Dibromobutane, 99%
CAS: 110-52-1 | C4H8Br2 | 215.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA161285000
Price (MYR)
1,118.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,118.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 4-Dibromobutane
Name Note99%
CAS110-52-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger