Veratrole, 99+%
Veratrole, 99+%
Veratrole, 99+%
Thermo Scientific Chemicals
Veratrole, 99+%
CAS: 91-16-7 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA161115000
Price (MYR)
556.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
556.00
BOT
Add to cart
Specifications
Chemical Name or MaterialVeratrole
Name Note99+%
CAS91-16-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement