1,3,5-Tribromobenzene, 98%
1,3,5-Tribromobenzene, 98%
1,3,5-Tribromobenzene, 98%
1,3,5-Tribromobenzene, 98%
Thermo Scientific Chemicals
1,3,5-Tribromobenzene, 98%
CAS: 626-39-1 | C6H3Br3 | 314.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA161081000
Price (MYR)
1,060.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,060.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 3, 5-Tribromobenzene
Name Note98%
CAS626-39-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning