3-Indolebutyric acid, 98%
3-Indolebutyric acid, 98%
3-Indolebutyric acid, 98%
3-Indolebutyric acid, 98%
Thermo Scientific Chemicals
3-Indolebutyric acid, 98%
CAS: 133-32-4 | C12H13NO2 | 203.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA160690250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Indolebutyric acid
CAS133-32-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0